missing translation for 'onlineSavingsMsg'
Learn More

Phenanthroline

Product Code. 18713018
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18713018 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18713018 Supplier Tocris Bioscience Supplier No. 6385/100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enhances hPSC differentiation into cranial placode cells

Phenanthroline is a enhances hPSC differentiation into cranial placode cells. Induces a 5-fold increase in cells expressing SIX1.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,10-Phenanthroline
CAS 66-71-7
Assay Percent Range >99%
Quantity 100 mg
Molecular Formula C12H8N2
Formula Weight Observed MW: 180.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.