missing translation for 'onlineSavingsMsg'
Learn More

Melford Phenanthroline

Product Code. 30018680
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018680 50 g 50g
30018679 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018680 Supplier Melford Supplier No. P3020050.0
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Forms a complex with ferrous ions.

Forms a complex with ferrous ions. It can be used as an indicator in oxidation-reduction systems in fitrating ferrous salts. Material passes test for redox indicator and passes test for iron determination.

Specifications

CAS 5144-89-8
Molecular Formula C12H8N2-H2O
Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.