missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene-d10, TRC

Product Code. 30417526
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30417526 Supplier Toronto Research Chemicals Supplier No. TRCP294801100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Phenanthrene D10
CAS 1517-22-2
Purity Grade Notes HPLC
Molecular Formula C14 2H10
Quantity 100 mg
Synonym Phenanthrene-1,2,3,4,5,6,7,8,9,10-d10, Phenanthrene-d10 (6CI,7CI,8CI,9CI), 1,2,3,4,5,6,7,8,9,10-Decadeuteriophenanthrene, Perdeuterated phenanthrene, Perdeuteriophenanthrene, Perdeuterophenanthrene, [2H10]Phenanthracene, Phenanthrene D10
InChI Formula InChI=1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMILES [2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c([2H])c3c([2H])c([2H])c([2H])c([2H])c23
IUPAC Name 1,2,3,4,5,6,7,8,9,10-decadeuteriophenanthrene
Molecular Weight (g/mol) 188.29
Formula Weight 188.141
Percent Purity >95
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.