missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene-9-boronic acid, 97%

Product Code. 15429287
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429287 5 g 5g
15419287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429287 Supplier Thermo Scientific Alfa Aesar Supplier No. H52546.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68572-87-2 | C14H11BO2 | 222.05 g/mol

Phenanthrene-9-boronic acid as a reactant is involved in the synthesis of large polycyclic aromatic hydrocarbons (PAHs) and electron rich benzo[g,h,i]perylenes. It is also used for preparation of FLAP protein inhibitors as anti-inflammatory agents and aminoisoquinolinones as PARP-2 selective inhibitors. As a reactant it is involved in homocoupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenanthrene-9-boronic acid as a reactant is involved in the synthesis of large polycyclic aromatic hydrocarbons (PAHs) and electron rich benzo[g,h,i]perylenes. It is also used for preparation of FLAP protein inhibitors as anti-inflammatory agents and aminoisoquinolinones as PARP-2 selective inhibitors. As a reactant it is involved in homocoupling reactions.

Solubility
Soluble in DMSO.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68572-87-2
Molecular Formula C14H11BO2
Molecular Weight (g/mol) 222.05
MDL Number MFCD00143524
InChI Key JCDAUYWOHOLVMH-UHFFFAOYSA-N
Synonym 9-phenanthreneboronic acid, 9-phenanthracenylboronic acid, phenanthrene-9-boronic acid, 9-phenanthrenylboronic acid, 9-phenanthrene boronic acid, 9-phenanthreneboronicacid, 9-phenanthrylboronic acid, boronic acid, 9-phenanthrenyl, phenanthracene-9-boronic acid, phenanthreneboronicacid
PubChem CID 11775704
IUPAC Name phenanthren-9-ylboronic acid
SMILES OB(O)C1=CC2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 254°C to 256°C
Quantity 5 g
Solubility Information Soluble in DMSO.
Formula Weight 222.05
Percent Purity 97%
Chemical Name or Material Phenanthrene-9-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.