missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Phenamil

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TRPP3 channel inhibitor (IC50 = 0.14 μM). Also inhibits epithelial Na+ channels (Kd = 0.4 nM for a high affinity site on the epithelial Na+ channel). Derivative of amiloride (Cat. No. 0890).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 401.83 |
| Purity | >99% |
| Molecular Weight (g/mol) | 401.826 |
| Chemical Name or Material | Phenamil |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction