missing translation for 'onlineSavingsMsg'
Learn More

Phenacetin, MedChemExpress

Product Code. 16402199
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16402199

Brand: Medchem Express HYB0476

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phenacetin (Acetophenetidin) is a non-opioid analgesic/antipyretic agent. Phenacetin is a selective COX-3 inhibitor. Phenacetin is used as probe of cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.

Phenacetin (Acetophenetidin) is a non-opioid analgesic/antipyretic agent. Phenacetin is a selective COX-3 inhibitor. Phenacetin is used as probe of cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.

Specifications

Chemical Name or Material Phenacetin
CAS 62-44-2
Color White
Purity Grade Notes Research
Molecular Formula C10H13NO2
Quantity 1 g
Synonym Acetophenetidin
Solubility Information DMSO : ≥ 100 mg/mL (557.97 mM) ∣H2O : 0.67 mg/mL (3.74 mM; Need ultrasonic)
SMILES CC(NC1=CC=C(OCC)C=C1)=O
Molecular Weight (g/mol) 179.22
Formula Weight 179.22
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.