missing translation for 'onlineSavingsMsg'
Learn More

Phellodendrine Chloride, TRC

Product Code. 30422975
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30422975 100 mg
30424689 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422975 Supplier Toronto Research Chemicals Supplier No. TRCP294565100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104112-82-5
Molecular Formula C20H24ClNO4
Molecular Weight (g/mol) 377.87
Synonym (7S-cis)5,8,13,13a-Tetrahydro-2,11-dihydroxy-3,10-dimethoxy-7-methyl-6H-Dibenzo[a,g]quinolizinium Chloride,
IUPAC Name (7S,13aS)-3,10-dimethoxy-7-methyl-6,8,13,13a-tetrahydro-5H-isoquinolino[3,2-a]isoquinolin-7-ium-2,11-diol;chloride
SMILES [Cl-].COc1cc2CC[N@@+]3(C)Cc4cc(OC)c(O)cc4C[C@H]3c2cc1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H23NO4.ClH/c1-21-5-4-12-8-19(24-2)18(23)10-15(12)16(21)6-13-7-17(22)20(25-3)9-14(13)11-21;/h7-10,16H,4-6,11H2,1-3H3,(H-,22,23);1H/t16-,21-;/m0./s1
Formula Weight 377.1394
Chemical Name or Material Phellodendrine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.