missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA133195
This item is not returnable.
View return policy
Description
PA1-33195 detects HIF Prolyl Hydroxylase 2 from mouse samples.
SYMPK is a nuclear protein that functions in the regulation of polyadenylation and promotes gene expression. The protein forms a high-molecular weight complex with components of the polyadenylation machinery. It is thought to serve as a scaffold for recruiting regulatory factors to the polyadenylation complex. It also participates in 3'-end maturation of histone mRNAs, which do not undergo polyadenylation. The protein also localizes to the cytoplasmic plaques of tight junctions in some cell types.This gene encodes a nuclear protein that functions in the regulation of polyadenylation and promotes gene expression. The protein forms a high-molecular weight complex with components of the polyadenylation machinery. It is thought to serve as a scaffold for recruiting regulatory factors to the polyadenylation complex. It also participates in 3'-end maturation of histone mRNAs, which do not undergo polyadenylation. The protein also localizes to the cytoplasmic plaques of tight junctions in some cell types.
Specifications
| PHD2 | |
| Polyclonal | |
| Unconjugated | |
| Egln1 | |
| AI503754; C1orf12; C1ORF12 antibody; ECYT3; egl nine homolog 1; egl nine-like protein 1; egl-9 family hypoxia inducible factor 1; egl-9 family hypoxia-inducible factor 1; EGLN 1 EGLN1; EGLN1; HALAH; HIF PH2; HIF prolyl hydroxylase 2; HIF-P4H 2; Hif-p4h-2; HIFPH2; HIF-PH2; HIF-prolyl hydroxylase 2; HPH2; HPH-2; Hypoxia-inducible factor prolyl hydroxylase 2; ORF13; P4H2; Phd2; PHD-2; PNAS-118; PNAS-137; Prolyl Hy; Prolyl hydroxylase domain-containing protein 2; SM20; SM-20; zinc finger MYND domain-containing protein 6; ZMYND6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112405, 308913, 54583 | |
| 4°C, do not freeze | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P59722, Q91YE3, Q9GZT9 | |
| Egln1 | |
| A synthetic peptide made to an internal portion of mouse PHD2/HIF Prolyl Hydroxylase 2 (between residues 300-400). | |
| 25 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction