missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PHD1 Recombinant Rabbit Monoclonal Antibody (K01_2Q15)

Product Code. 30828914
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828914 50 μL
30828592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828914 Supplier Invitrogen™ Supplier No. MA565706
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: SSEPLEPEPG RARM.

Prolyl hydroxylase domain proteins HIF PHD1, HIF PHD2 and HIF PHD3 (known as PHD1, PHD2 and PHD3 in rodents, respectively) can hydroxylate HIF-alpha subunits. Hypoxia-inducible factor (HIF) is a transcriptional regulator important in several aspects of oxygen homeostasis. The prolyl hydroxylases catalyze the posttranslational formation of 4-hydroxyproline in HIF-alpha proteins. HIF PHD1, which is widely expressed, with highest levels of expression in testis, functions as a cellular oxygen sensor and is important in cell growth regulation. HIF PHD1 can localize to the nucleus or the cytoplasm and is also detected in hormone responsive tissues, such as normal and cancerous mammary, ovarian and prostate epithelium. HIF PHD1 is encoded by EGLN2, which maps to chromosome 19q13. 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHD1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2Q15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene EGLN2
Gene Accession No. Q6AYU4, Q96KS0
Gene Alias 0610011A13Rik; C85656; egl nine homolog 2; egl-9 family hypoxia inducible factor 2; egl-9 family hypoxia-inducible factor 2; Egln2; EIT6; EIT-6; Estrogen-induced tag 6; falkor; HIF-1alpha prolyl-4-hydroxylase-1; HIF-P4H 1; Hif-p4h-1; HIFPH1; HIF-PH1; HIF-prolyl hydroxylase 1; HPH-1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 1; Ier4; immediate early response 4; P4H1; Phd1; PHD-1; Prolyl hydroxylase domain-containing protein 1; SM-20
Gene Symbols EGLN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PHD1/prolyl hydroxylase (aa 21-34).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112398, 308457
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.