missing translation for 'onlineSavingsMsg'
Learn More

PHC3 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18416675 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416675 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416675 Supplier Novus Biologicals Supplier No. NBP282020IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PHC3 Polyclonal antibody specifically detects PHC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHC3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias DKFZp313K1221, early development regulator 3, Early development regulatory protein 3, EDR3FLJ12729, FLJ12967, Homolog of polyhomeotic 3, HPH3, MGC88144, PH3, polyhomeotic homolog 3 (Drosophila), polyhomeotic like 3, polyhomeotic like 3 (Drosophila), polyhomeotic-like protein 3
Host Species Rabbit
Immunogen PHC3 antibody was raised against a 19 amino acid peptide near the amino terminus of human PHC3. The immunogen is located within amino acids 240 - 290 of PHC3. Amino Acid Squence: KISQRDPSPESNKKGESPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 80012
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.