missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12867-1-AP-150UL
This item is not returnable.
View return policy
Description
In Drosophila melanogaster, the ′Polycomb′ group (PcG) of genes are part of a cellular memory system that is responsible for the stable inheritance of gene activity. PcG proteins form a large multimeric, chromatin-associated protein complex. The protein encoded by this gene has homology to the Drosophila PcG protein ′polyhomeotic′ (Ph) and is known to heterodimerize with EDR1 and colocalize with BMI1 in interphase nuclei of human cells. The specific function in human cells has not yet been determined. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| PHC2 | |
| Polyclonal | |
| Unconjugated | |
| PHC2 | |
| EDR2, HPH2, PH2, PHC2, Polyhomeotic like protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1912, 313038, 54383 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IXK0, Q9QWH1 | |
| PHC2 | |
| PHC2 Fusion Protein Ag3507 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction