Learn More
Invitrogen™ PHC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113941
Description
Immunogen sequence: GGSGRPTGPQ ISVYSGIPDR QTVQVIQQAL HRQPSTAAQY LQQMYAAQQQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
In Drosophila melanogaster, the 'Polycomb' group (PcG) of genes are part of a cellular memory system that is responsible for the stable inheritance of gene activity. PcG proteins form a large multimeric, chromatin-associated protein complex. The protein encoded by this gene has homology to the Drosophila PcG protein 'polyhomeotic' (Ph) and is known to heterodimerize with EDR1 and colocalize with BMI1 in interphase nuclei of human cells. The specific function in human cells has not yet been determined. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| PHC2 | |
| Polyclonal | |
| Unconjugated | |
| Phc2 | |
| A3galt2; AA415044; D130050K19Rik; D4Ertd810e; early development regulator 2; early development regulator 2 (homolog of polyhomeotic 2); early development regulatory protein 2; Edr2; hPH2; mPH2; p36; Ph2; Phc2; polyhomeotic 2; polyhomeotic 2 protein; polyhomeotic homolog 2; polyhomeotic homolog 2 (Drosophila); polyhomeotic-like 2; polyhomeotic-like 2 (Drosophila); polyhomeotic-like protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1912 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IXK0 | |
| Phc2 | |
| Synthetic peptide directed towards the N terminal region of human PHC2 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.