missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PhastGel™ Buffer Strips
Shop All Cytiva Products
Click to view available options
Quantity:
2 Pack
20 Strips
For Use With (Application):
Gel Electrophoresis
Product Type:
Buffer strip holder
PhastGel Buffer strips-SDS
PhastGel Buffer strips-native
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Description
- PhastGel Buffer Strips are available in three types – PhastGel DNA, PhastGel SDS, and PhastGel Native
- Select from homogeneous and gradient polyacrylamide gels.
- Save time and improve reproducibility with precast gels, buffer strips, and staining kits.
- Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
Specifications
Specifications
| Buffer | 0.2M Tricine, 0.2M Tris, 0.55% SDS, pH 8.1 |
| Color | Colorless |
| For Use With (Equipment) | PhastSystem Automated Electrophoresis Unit |
| Product Type | PhastGel Buffer strips-SDS |
| Form | Solid |
| Formulation | N-(Tri(Hydroxymethyl)Methyl)Glycine 1 to 5%, Trometamol 1 to 5% |
| For Use With (Application) | Gel Electrophoresis |
| Quantity | 20 Strips |
| Storage Requirements | 4° to 8°C |
| Sufficient For | 10 PhastGel™ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction