missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva PhastGel™ Buffer Strips
Shop All Cytiva Products
Click to view available options
Quantity:
2 Pack
20 Strips
For Use With (Application):
Gel Electrophoresis
Product Type:
Buffer strip holder
PhastGel Buffer strips-SDS
PhastGel Buffer strips-native
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Beskrivelse
- Ready made buffer strips eliminate the need to prepare electrophoresis buffers
- Supplied as 10X pairs - for both cathode and anode
- Choice of Native or Denaturing (SDS) buffer systems
Tekniske data
Tekniske data
| Buffer | 0.88M L-alanine, 0.25M Tris, pH 8.8 |
| Color | Colorless |
| For Use With (Equipment) | PhastSystem Automated Electrophoresis Unit |
| Product Type | PhastGel Buffer strips-native |
| Form | Solid |
| Formulation | Trometamol 1 to 5% |
| For Use With (Application) | Gel Electrophoresis |
| Quantity | 20 Strips |
| Storage Requirements | 4° to 8°C |
| Sufficient For | 10 PhastGel™ |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion