Learn More
Carl Zeiss™ Phase Telescope
Descripción
An innovative inverted microscope with superior optical performance across a wide range of functions.
- An inverted microscope for cell and molecular biology
- Ideal for examining living cells
- Easy to operate, robust, ergonomic design and features quality optics
- Efficient illumination: halogen lamp with more than an 800hr life
- Two effective techniques for thin specimens: Brightfield and phase contrast
- Two effective techniques for thick specimens: VAREL (variable relief contrast) and PlasDIC (differential interference contrast)
- Exceptionally high fluorescence standards using the Axiovert 40 CFL
- Flexible upgrading for future demands
Specifications
Specifications
| Type | Phase Telescope |
| For Use With (Equipment) | Axiovert |
| Quantity | 1 Ea. |