Learn More
Motic™ Phase Slider, Empty
Product Code. 11712358
Change view
Click to view available options
Type:
Phase Slider, Empty
Unit Size:
Each
Product Code. Type unitSize
11712358 Phase Slider, Empty Each
1 options
Product Code. 11712358
Brand: Motic™ 1101009700092
Only null left
Add to basket
Add to basket
Use phase contrast to improve visualization of transparent specimens. Motic™ Phase slider, Empty is a centerable, three-position phase slider.
  • Boost contrast and detail discrimination without staining or fixing, thus permitting live cell study.
  • Must be matched and aligned to the objective phase plate.
Specifications
Type Phase Slider, Empty