Learn More
Medline Scientific™ Phase Contrast Kit
Click to view available options
Magnification Power:
10x/ - 20x/ - 40x/ - 100x(oil)
Unit Size:
Each
Description
- Positive turret condenser
- With iris diaphragm
- 4 pos. phase planachromatic objectives 10x/ - 20x/ - 40x/ - 100x(oil)
- Centring telescope
- Green and yellow filters
Specifications
Specifications
| Type | Phase Contrast Kit |
| Description | Positive turret condenser |
| Includes | Green and yellow filters |
| Magnification Power | 10x/ - 20x/ - 40x/ - 100x(oil) |