missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Some cyanine-type dyes (PE-Cy5, PerCP-Cy5.5, PE-Cy7, APC-Cy7, APC-H7, and others) and non-cyanine tandem dyes (eg, PE-CF594) are known to bind to human and mouse monocytes, macrophages, and other leukocyte subsets, though the exact mechanism is not clear. BD Pharmingen™ MonoBlock™ Leukocyte Staining Buffer reduces this nonspecific background staining, while not impacting specific surface staining or cell viability. This buffer is added directly to leukocytes or added to fluorescent reagent cocktails prior to staining leukocytes. This reagent may be complemented by others including i) BD Horizon™ Brilliant Stain Buffer, which mitigates dye-dye interactions amongst BD Horizon™ Brilliant fluorochromes, and ii) BD Fc Block™ reagents. The latter can reduce potential nonspecific staining caused by fluorescent antibodies that bind to Fc receptors expressed by human and mouse cells. For optimal and reproducible surface staining results, BD Pharmingen™ MonoBlock™ Leukocyte Staining Buffer should be used with cyanine-like and other tandem dye-conjugated reagents that can potentially stain cells nonspecifically. Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Specifications
Specifications
| Color | Colorless to Yellow |
| Format | Aqueous Liquid |
| For Use With (Application) | Flow Cytometry |
| For Use With (Equipment) | Flow Cytometer |
| Quantity | 100 Tests |
| Grade | Research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?