missing translation for 'onlineSavingsMsg'
Learn More

Pharmco 18.93 LT (5 GAL) SDA 30-190-proof Poly-W-Reike-Spout, Pharmco

Product Code. 30286184
Change view
Click to view available options
Quantity:
5 Gallon
Unit Size:
18.93 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30286184 5 Gallon 18.93 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 30286184 Supplier Pharmco Supplier No. 12330190PL05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Specifications

CAS Ethyl alcohol 64-17-5 84.21 - 90.88 Chemical name CAS number % Methanol 67-56-1 8.89 - 9.12 Water 7732-18-5 0 - 6.90
Quantity 5 Gallon
Packaging 5 Gallon Poly Pail
Boiling Point 77.1 - 78.4
Chemical Name or Material Specially Denatured Alcohol (SDA) 30 190 Proof
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.