missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pharmco 16 LT Butyl-alcohol N-1-Butanol NF-USP-Amber-Glass (4 x 4 LT), Pharmco
Description
6 LT Butyl-alcohol N-1-Butanol NF-USP-Amber-Glass (4 x 4 LT)
Specifications
Specifications
| CAS | 71-36-3 |
| Quantity | 4x1 Fluorinated Poly Gallon Case |
| Packaging | 4x1 Fluorinated Poly Gallon Case |
| Boiling Point | 119°C |
| Grade | USP |
| Chemical Name or Material | n-Butyl Alcohol, USP/NF |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?