Learn More
Spectrum Labs™ PharmaPure™ Tubing
Description
- Nontoxic and nonhemolytic with excellent biocompatability
- Low spallation
- Low extractables and low gas permeability
- Superior flex life
- Meets FDA and USP Class VI criteria
- Developed for pharmaceutical, biotechnology, and laboratory applications
Specifications
Specifications
| Diameter (English) Inner | 0.31 in. |
| Description | Low spallation, biologically compatible |
| For Use With | 3/8 in. hose barb |
| Material | PharmaPure |