missing translation for 'onlineSavingsMsg'
Learn More

Köttermann Pharmacy Fume Hood without Media

Product Code. 17322189
Click to view available options
Configuration Options:
No Ventilation Fan
Ventilation Fan
Plug Type:
GB Plug
Plug Type D (Old British Plug)
Schuko Plug
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17322189

Brand: Köttermann 45100026

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Pharmacy Fume Hood without Media is designed for the handling of hazardous substances in pharmacies and is ideally suited for working with dusty preparations or volatile chemicals.

  • For working safely with dust-forming preparations and volatile chemicals
  • Long-lasting steel construction with one-piece polyacrylic sash
  • Ergonomic features to improve workflow
  • Low, compact design for small laboratories

Specifications

Configuration Options Ventilation Fan
Dimensions (L x W x H) 640 x 900 x 1350 mm
Plug Type GB Plug
Voltage 230 V
Weight (Metric) 115 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.