missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ Phagotest (CE/IVD)

Product Code. 10664775
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10664775

Brand: BD Biosciences™ 341060

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent kit for the quantitative determination of the phagocytic activity of monocytes and granulocytes in heparinized human whole blood.

  • This reagent kit faciltates the quantitative determination of leukocyte phagocytosis in heparinized whole blood.
  • It contains fluorescein (FITC)-labelled opsonized E.coli bacteria and necessary reagents.
  • It measures the overall percentage of monocytes and granulocytes showing phagocytosis in general (ingestion of one or more bacteria per cell) and the individual cellular phagocytic activity number of bacteria per cell.
  • Provides accurate quantitative analysis suitable for qualitiy comparisons.
  • Detailed instructions provided result from specific experience and precise validation assays.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Instamed-Salts, Stabilized and Opsonized FITC-labeled E.coli Suspension, Quenching Solution, Lysing Solution, DNA Staining Solution
Includes 1 L Instamed-Salts, 2 mL Stabilized and Opsonized FITC-labeled E.coli Suspension, 10 mL Quenching Solution, 20 mL Lysing Solution, 20 mL DNA Staining Solution
No. of Tests 100 Tests
Quantity 100 Tests
Stability The reagents are stable for the period shown on the packaging label, when stored as described.
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1A
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H311 + H331-Toxic in contact with skin or if inhaled.
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.,Suspected of causing genetic defects.
  • H350-May cause cancer.,May cause cancer.
  • H373-May cause damage to organs.,May cause damage to organs.
  • H371-May cause damage to organs.,May cause damage to organs.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P310-Immediately call a POISON CENTER/doctor/physician.
  • P501-Dispose of contents/container to an approved waste disposal plant..,Dispose of contents/container to an approved waste disposal plant..
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.,IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : modified alkyl carbomixture of 5-Chloro-Formaldehydediethylene glycolmethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.