missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Phago'Wash DA

Product Code. p-200000170
Click to view available options
Volume (Metric):
5 L
Unit Size:
10 L
This item is not returnable. View return policy

Product Code. 16545531

Brand: Christeyns™ 70440

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phago'Wash DA is a non-foaming alkaline liquid detergent for cleaning laboratory equipment and glassware.

  • Can be used on surfaces that come into contact with foodstuffs, after rinsing with drinking water
  • Powerful alkaline detergent
  • Free of NTA, EDTA or phosphates

Specifications

Product Type Alkaline Cleaning Agent
Color-coded No
Form Concentrated Liquid
pH 11.5 to 12.5
Product Line Cleanware
Scent Unscented
Sterility Non-sterile
Quantity 2 Cans
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • PHAGO'WASH DA
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,ethanol, sodium metasilicate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.