missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Christeyns™ Phago'Spray DASR 70
Click to view available options
Scent:
Fresh Scent
Unscented
Size:
750 mL
5 L
Unit Size:
Pack of 2
Pack of 6
Description
- Can be used in application on a single-use cloth for the rapid disinfection of small furniture, trolleys, external surfaces of non-invasive medical devices (operating table, operating light, monitors, cables and connectors, stethoscopes, reflex, blood pressure monitors) and small surfaces close to the patient
- Can be used, without rinsing, on surfaces that may be in direct contact with foodstuffs (table, worktop, trays, meal trolley, slicers, choppers, knives, etc.)
- Broad antimicrobial spectrum from 30 sec.
- Food contact, without rinsing
- No streaks or residue on drying
- Ecocertified
Specifications
Specifications
| Product Type | Alcohol-based Surface Disinfectant |
| Color-coded | No |
| Form | Clear Liquid |
| pH | Neutral |
| Scent | Unscented |
| Sterility | Non-sterile |
| Quantity | 2 Cans |
| Size | 5 L |
Safety and Handling
Product Identifier
- phago'spray DM
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : ethanol, didecyldimethylammonium chloride, N-(3-aminopropyl)-N-dodecylpropane-1,3-diamine, ethane-1,
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction