missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Christeyns™ Phago'Spray DASR 70
Description
- Can be used in application on a single-use cloth for the rapid disinfection of small furniture, trolleys, external surfaces of non-invasive medical devices (operating table, operating light, monitors, cables and connectors, stethoscopes, reflex, blood pressure monitors) and small surfaces close to the patient
- Can be used, without rinsing, on surfaces that may be in direct contact with foodstuffs (table, worktop, trays, meal trolley, slicers, choppers, knives, etc.)
- Broad antimicrobial spectrum from 30 sec.
- Food contact, without rinsing
- No streaks or residue on drying
- Ecocertified
Specifications
Specifications
| Product Type | Alcohol-based Surface Disinfectant |
| Color-coded | No |
| Form | Clear Liquid |
| pH | Neutral |
| Scent | Fresh Scent |
| Sterility | Non-sterile |
| Quantity | 6 Bottles |
| Size | 750 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?