missing translation for 'onlineSavingsMsg'
Learn More

PHACTR3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£143.00 - £251.00

Specifications

Antigen PHACTR3
Dilution Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18671709
View Documents
Novus Biologicals
NBP3-12697-100ul
100 μg
£251.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18632518
View Documents
Novus Biologicals
NBP3-12697-50ul
50 μg
£143.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PHACTR3 Polyclonal antibody specifically detects PHACTR3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

PHACTR3
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
C20orf101, chromosome 20 open reading frame 101, H17739, MGC117178, phosphatase and actin regulator 3, scaffold-associated PP1 inhibiting protein, Scaffold-associated PP1-inhibiting protein, SCAPIN1, Scapinin
Produced in rabbits immunized with E. coli-derived Human PHACTR3 fragment
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Purified
RUO
PBS, pH7.0
116154
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.