missing translation for 'onlineSavingsMsg'
Learn More

PHA Reagent Grade

Product Code. 13215729
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13215729 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13215729 Supplier Thermo Scientific Oxoid Supplier No. R30852701
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Facilitate cytogenic studies of chromosomes by stimulating the mitotic division of lymphocytes with this reagent.

Facilitate cytogenic studies of chromosomes by stimulating the mitotic division of lymphocytes maintained in a cell culture system with Thermo Scientific™ Remel™ PHA Reagent Grade. Phytohaemagglutinin (PHA) is universally effective and convenient stimulant of lymphocytes from many animal species. The widespread popularity of peripheral blood culture as a means of chromosome analysis has been largely dependent on this property.

Use Remel Phytohaemagglutinin to stimulate mitotic division of lymphocytes maintained in a cell culture system and facilitate cytogenic studies of chromosomes.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Phytohaemagglutinin (Reagent Grade)
CE Marker Yes
Type PHA
Includes 1 bottle Phytohaemagglutinins (reagent grade)
Quantity 5 mL
Species Animal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.