missing translation for 'onlineSavingsMsg'
Learn More

PHA-680632, MedChemExpress

Product Code. 16486287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486287 5 mg 5mg
16459976 50 mg 50mg
16454676 100 mg 100mg
16443568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486287 Supplier Medchem Express Supplier No. HY101785mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PHA-680632 is an aurora kinase inhibitor with IC50s of 27, 135 and 120 nM for aurora A, B and C, respectively.

PHA-680632 is an aurora kinase inhibitor with IC50s of 27, 135 and 120 nM for aurora A, B and C, respectively.

Specifications

Chemical Name or Material PHA-680632
Physical Form Solid
CAS 398493-79-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C28H35N7O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 30 mg/mL (59.81 mM)
SMILES O=C(N1CC2=C(NN=C2NC(C3=CC=C(N4CCN(C)CC4)C=C3)=O)C1)NC5=C(C=CC=C5CC)CC
Molecular Weight (g/mol) 501.62
Formula Weight 501.62
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.