missing translation for 'onlineSavingsMsg'
Learn More

PHA-665752, MedChemExpress

Product Code. 16484398
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484398 10 mg 10mg
16480717 50 mg 50mg
16445296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16484398 Supplier Medchem Express Supplier No. HY11107
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PHA-665752 is a selective, ATP-competitive, and active-site inhibitor of the catalytic activity of c-Met kinase (Ki=4 nM; IC50=9 nM). PHA-665752 exhibits >50-fold selectivity for c-Met compared with a panel of diverse tyrosine and serine-threonine kinases. PHA-665752 induces apoptosis and cell cycle arrest, and exhibits cytoreductive antitumor activity.

PHA-665752 is a selective, ATP-competitive, and active-site inhibitor of the catalytic activity of c-Met kinase (Ki=4 nM

Specifications

Chemical Name or Material PHA-665752
CAS 477575-56-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H34Cl2N4O4S
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (77.93 mM; Need ultrasonic)
SMILES O=C(C1=C(NC(/C=C2C(NC3=C\2C=C(C=C3)S(=O)(CC4=C(C=CC=C4Cl)Cl)=O)=O)=C1C)C)N5[C@H](CCC5)CN6CCCC6
Molecular Weight (g/mol) 641.61
Formula Weight 641.61
Percent Purity 96.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.