missing translation for 'onlineSavingsMsg'
Learn More

PHA-665752, MedChemExpress

Product Code. 16445296
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16445296

Brand: Medchem Express HY11107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PHA-665752 is a selective, ATP-competitive, and active-site inhibitor of the catalytic activity of c-Met kinase (Ki=4 nM; IC50=9 nM). PHA-665752 exhibits >50-fold selectivity for c-Met compared with a panel of diverse tyrosine and serine-threonine kinases. PHA-665752 induces apoptosis and cell cycle arrest, and exhibits cytoreductive antitumor activity.

PHA-665752 is a selective, ATP-competitive, and active-site inhibitor of the catalytic activity of c-Met kinase (Ki=4 nM

Specifications

Chemical Name or Material PHA-665752
CAS 477575-56-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H34Cl2N4O4S
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (77.93 mM; Need ultrasonic)
SMILES O=C(C1=C(NC(/C=C2C(NC3=C\2C=C(C=C3)S(=O)(CC4=C(C=CC=C4Cl)Cl)=O)=O)=C1C)C)N5[C@H](CCC5)CN6CCCC6
Molecular Weight (g/mol) 641.61
Formula Weight 641.61
Percent Purity 96.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.