missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ PICCOLO™ plus pH/Temperature Tester with 6.3 in. Probe

Product Code. 12903227
Click to view available options
Range (pH):
1 to 13
This item is not returnable. View return policy

Product Code. 12903227

Brand: Hanna Instruments™ PICCOLOPLUS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Delivers a strong signal to the meter. Hanna Instruments™ PICCOLO™ plus pH/Temperature Tester with 6.3 in. Probe has a 4-in-1 amplified electrode. The interchangeable electrode is inexpensive, rugged and houses the pH sensor, reference system, temperature sensor and amplifier module.

The PICCOLO™ plus is a lightweight digital tester that is easy to use with a replaceable probe. Conventional pH meters are susceptible to the weak, high impedance signal which makes the electrode, connector, cable and meter vulnerable to noise, humidity and dirty environments. PICCOLO™ Plus has overcome these problems with a revolutionary interchangeable amplified electrode.

  • Replaceable 6.3 in. Pre-amplified pH Electrode
  • ±0.01 pH Accuracy
  • Temperature readout
  • Automatic Temperature Compensation
  • Easy two-point manual calibration
  • 100 hour battery life
  • Simple to use
  • Economical

Specifications

Includes HI1295 electrode, pH 4.01 and pH 7.01 buffer solution sachets, calibration screwdriver, batteries, rugged carrying case and instructions
Accuracy (Temperature) ±1°C
Accuracy (pH) ±0.01
Calibration Manual
Range (pH) 1 to 13
Resolution (pH) 0.01
Calibration Points 2
Temperature Range (Metric) 0°C to 70°C
Temperature Range (English) 32°F to 158°F
Resolution (Temperature) 0.1°C
Dimensions (L x W x H) 265 x 29 x 15 mm (10.4 x 1.1 x 0.6 in.)
Weight (Metric) 70 g
Weight (English) 2.5 oz.
Warranty 1 Year (6 Months Probe)
Probe Type HI1295 amplified electrode with temperature sensor
Relative Humidity Range 95% max
Temperature Compensation Automatic from 0 to 70°C (32 to 158°F)
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Operating 0°C to 50°C
Battery Type 3 x 1.5V
Type pH Temperature Stick
Battery Life 100 hr.
Length (English) 6.3 in. Probe
Length (Metric) 160 mm Probe
Width (Metric) 265 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.