missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH and Redox Electrode for Mounting Bracket

Maintenance-free

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11754809

  • £166.42 / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

  • Supplied with 3m cable and BNC plug
  • Available in standard and double junction type
  • Double junction electrode is suitable for media containing heavy metals, sulphides, substances and Organic compounds
Specifications

Specifications

BNC Cable
3 m
±2000
Double Junction
Redox Electrode
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers