missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Blue Litmus Paper

Product Code. 10190001 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
8 to 5
Unit Size:
Pack of 100
Product Code. pH Range unitSize
10190001 8 to 5 Pack of 100
1 options
This item is not returnable. View return policy

Product Code. 10190001

Brand: Macherey-Nagel™ 91126

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides a general test for acidic or alkaline reactions. Macherey Nagel™ Blue Litmus Paper is comprised of completely impregnated indicator paper.

Provides a general test for acidic or alkaline reactions. Macherey Nagel™ Blue Litmus Paper is comprised of completely impregnated indicator paper.

  • Color change from blue to red
  • Store in a cool, dry place
  • Avoid exposure to sunlight and moisture
  • Serves in neutralization reactions to check whether the change from acid to alkaline or vice versa has taken place

Specifications

Color Indicator Blue to Red
pH Range 8 to 5
Format Strip
For Use With (Application) Determination of acidic and alkaline reaction
Quantity 100 Pack
Type pH Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.