missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals pH Indicator solution, universal (in isopropanol), pure

Product Code. 10209660 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 10209660

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material pH Indicator solution, universal (in isopropanol)
CAS 67-63-0, 7732-18-5
MDL Number MFCD00148741
Grade Extra Pure
Packaging EcoSafPak™ Plastic Bottle
Vapor Pressure Vapor Pressure: 25mmHg at 20°C
Visual Transition Interval pH 4.0 (red), pH 4.5 (orange-red), pH 5.0 (orange), pH 5.5 (orange-yellow), pH 6.0 (yellow), pH 6.5 (yellow-green), pH 7.0 (green), pH 7.5 (green, slightly blue), pH 8.0 (green-blue), pH 8.5 (blue-green), pH 9.0 (blue)
Density 1.3
Quantity 500 mL
Physical Form Solution
compliance-icons compliance-icons
Product Identifier
  • pH Indicator solution, universal (in isopropanol)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Isopropyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.