missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH Electrode

Product Code. 11739738
Click to view available options
Length (Metric):
102 mm
115 mm
120 mm
152 mm
225 mm
Unit Size:
Each

Product Code. 11739738

This item requires a price update prior to being ordered.

Request a Price for this item here.

This unique maintenance free solid electrolyte pH electrode for measurement up to 110 ° C and 10 bar, is ideal for industrial applications.

  • High temperature pH electrode
  • Maintenance-free pH electrode with 3/4 "MNPT thread for line installation up to 7 bar and 100 ° C
  • Body in Kynar for better chemical resistance with double ceramic junction insensitive to pollution
  • Supplied with 3 m cable and BNC plug

Specifications

Description High temperature pH electrode
Includes 3 m cable and BNC connector
Length (Metric) 152 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.