Learn More
Cytiva pH Electrode with Cell and Holder
For continuous pH measurement in a liquid chromatography system
Brand:  Cytiva 18-1134-84
Code : GX
Additional Details : Weight : 1.16000kg
Product Code. 11340262
  • £2017.00 / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Includes: cell and holder
Description
Description
  • Provides temperature-compensated pH measurements
  • Made from FFKM, Ti and glass
Specifications
Specifications
For continuous pH measurement
pH Electrode
0 to 14
Liquid Chromatography
cell and holder