Learn More
Fisherbrand™ pH Electrode
Product Code. 11705628
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
Product Code. Color Unit Size
11705628 Blue Each
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11705628 Supplier Fisherbrand™ Supplier No. 5732526B010BB
  • ONLINE EXCLUSIVE PRICE
    £42.15
    £30.95 / Pack of 1
    Save £11.20
    26% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Offered in a variety of styles including different reference solutions and reference chambers. Fisherbrand™ pH Electrode feature a refillable outer reference reservoir that can be filled with any electrolyte compatible with the sample, including those fills recommended for nonaqueous measurements.
  • Double junction reference is recommended for samples containing organic compounds, proteins, heavy metals or compounds that interact with silver (e.g. sulfide, bromide, iodide)
  • All electrodes are available with a detachable lead or with a cable/connector attached
  • Designed for all existing Brinkmann, Metrohm and Schott titration systems
  • The electrode body is constructed with a glass ground joint that will fit their titration vessels
  • Ionics and Tytronics titrators can be fitted with this same electrode without the ground joint
  • Either design can be ordered depending on the titration unit
Specifications
Color Blue
Temperature (Metric) Range 0°C to 80°C