missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Mercury free buffers at 25°C (pH4 and pH10 are colored) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.
Specifications
Specifications
| Color | Undesignated |
| pH | 5 |
| Quantity | 10 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
