Learn More
pH Buffer Solution Kit pH 7.01, 10.01, Hanna Instruments™
Click to view available options
Quantity:
10 mL
Unit Size:
Pack of 10
Specifications
Specifications
| pH | 7.01, 10.01 |
| Quantity | 10 mL |
| Packaging | 10Pack |
| Traceability to NIST | Yes |