missing translation for 'onlineSavingsMsg'
Learn More

pH 9.00 Buffer Solution at 25°C, Reagecon

Tested, certified, NIST traceable, and shelf-stable buffer solution.

Brand:  Reagecon 509025

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11464559

  • £97.25 / 5 L
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • pH Buffer Reference Standard, pH Buffer Reference Material
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications

Specifications

Accuracy ± 0.01 pH units
Liquid
HDPE Bottle GL45
9.00 ±0.01
5 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers