missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 7.00 Technical Color-Coded Buffer Solution at 25°C, Reagecon
Shop All Reagecon Products

Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- NIST traceable
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
| Color | Yellow |
| Description | Accuracy ± 0.01 pH units |
| pH | 7.00 ±0.02 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | HDPE Twin Neck Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction