missing translation for 'onlineSavingsMsg'
Learn More

pH 7.00 Recal Color-Coded Buffer Solution at 25°C, Reagecon
BIOPHARMA

Product Code. 11414649 Shop All Reagecon Products
Click to view available options
Quantity:
6 x 90 mL
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 11414649

Brand: Reagecon 07C65

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Easy-to-use, tested, certified, NIST traceable, and shelf-stable, color-coded buffer solution.

  • pH Buffer Reference Standard, pH Buffer Reference Material
  • Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
  • NIST traceable

Specifications

Color Yellow
Description Accuracy ± 0.01 pH units
pH 7.00 ±0.01
Physical Form Liquid
Quantity 6 x 90 mL
Packaging HDPE Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.