missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 4.6 Acetate Buffer Solution According to European Pharmacopoeia (EP), Reagecon
Shop All Reagecon Products

Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
- Buffer Solution according to European pharmacopoeia (EP)
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
| Description | Accuracy ± 0.02 pH units |
| For Use With (Application) | Product monographs |
| pH | 4.6 ±0.02 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | HDPE Bottle GL45 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction