Learn More
pH 4.00 Phthalate-Free Buffer Solution at 20°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Brand: Reagecon CC1040
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Phthalate-free
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Accuracy ± 0.01 pH units | |
Liquid | |
HDPE Bottle GL45 |
4.00 ±0.01 | |
1 L |
Safety and Handling
- BUFFER SOLUTION pH 4.00 ± 0.01 @ 20°C
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : citric acid monohydrate