missing translation for 'onlineSavingsMsg'
Learn More

PH-002, MedChemExpress

Product Code. 16487417
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487417 5 mg 5mg
16473206 25 mg 25mg
16450997 50 mg 50mg
16444708 10 mg 10mg
16425536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487417 Supplier Medchem Express Supplier No. HY112798

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PH-002 is an inhibitor of apolipoprotein (apo) E4 intramolecular domain interaction in neuronal cells that could rescue impairments of mitochondrial motility and neurite outgrowth.

PH-002 is an inhibitor of apolipoprotein (apo) E4 intramolecular domain interaction in neuronal cells that could rescue impairments of mitochondrial motility and neurite outgrowth.

Specifications

Chemical Name or Material PH-002
Physical Form Solid
CAS 1311174-68-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C27H33N5O4
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 75 mg/mL (152.57 mM)
SMILES O=C(N1CCN(CC2=CC=C(NC(CC3=NN(C)C(C4=C3C=CC=C4)=O)=O)C=C2)CC1)OC(C)(C)C
Molecular Weight (g/mol) 491.58
Formula Weight 491.58
Percent Purity 98.84%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.