missing translation for 'onlineSavingsMsg'
Learn More

PGS-IN-1, MedChemExpress

Product Code. p-200100413
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16483438

Brand: Medchem Express HY101587

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PGS-IN-1 is a potent inhibitor of prostaglandin synthetase (PGS) with an IC50 of 0.28 μM; also inhibits 5-lipoxygenase with an IC50 of 1.05 μM.

PGS-IN-1 is a potent inhibitor of prostaglandin synthetase (PGS) with an IC50 of 0.28 μM

Specifications

Chemical Name or Material PGS-IN-1
CAS 102271-49-8
Color White
Purity Grade Notes Research
Molecular Formula C19H26O3
Quantity 10 mg
Synonym KME-4
SMILES O=C1OCC/C1=C\C2=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C2
Molecular Weight (g/mol) 302.41
Formula Weight 302.41
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.