missing translation for 'onlineSavingsMsg'
Learn More

PGS-IN-1, MedChemExpress

Product Code. 16483438
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483438 10 mg 10mg
16460426 20 mg 20mg
16436167 5 mg 5mg
16400799 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483438 Supplier Medchem Express Supplier No. HY101587
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PGS-IN-1 is a potent inhibitor of prostaglandin synthetase (PGS) with an IC50 of 0.28 μM; also inhibits 5-lipoxygenase with an IC50 of 1.05 μM.

PGS-IN-1 is a potent inhibitor of prostaglandin synthetase (PGS) with an IC50 of 0.28 μM

Specifications

Chemical Name or Material PGS-IN-1
CAS 102271-49-8
Color White
Purity Grade Notes Research
Molecular Formula C19H26O3
Quantity 10 mg
Synonym KME-4
SMILES O=C1OCC/C1=C\C2=CC(C(C)(C)C)=C(O)C(C(C)(C)C)=C2
Molecular Weight (g/mol) 302.41
Formula Weight 302.41
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.