missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGRP-S Monoclonal Antibody (188C424)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA141044
This item is not returnable.
View return policy
Description
Suggested positive control: mouse spleen lysate or protein. Immunogen sequence: YVLKGHRDVQ RTLSPG.
The primary immune recognition is based on structures common among invading pathogens. Bacterial surface molecules, such as lipopolysaccharide (LPS) and peptidoglycan (PGN), are known to elicit immune reactions ranging from cytokine release to fever. Recently, a family of proteins called peptidoglycan recognition protein (PGRP) has been identified in mouse and human that binds to peptidoglycans expressed on Gram-positive bacteria. Peptidoglycan (PGN) is an essential cell wall component of virtually all bacteria (1,2) and, thus, it is an excellent target for recognition by the eukaryotic innate immune system. The PGRPs (PGRP-L, PGRP-S, PGRP-Ia, and PGRP-Ib) define a new family of human pattern recognition molecules (3). PGRP-L is primarily expressed in the liver. Although liver is not considered a primary immune organ, liver participates in host defenses by producing acute phase proteins (by hepatocytes) in response to infections and by clearing microorganisms from blood (4). PGRP-S is present in neutrophils and inhibits growth of Gram-positive bacteria and, therefore, may function as a neutrophil antibacterial protein (5). However, PGRP-S may have another, as yet unidentified function because in humans it is expressed in the bone marrow 50-100 times higher than in neutrophils.
Specifications
| PGRP-S | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.05% sodium azide | |
| O75594, O88593 | |
| PGLYRP1 | |
| KLH-conjugated synthetic peptide corresponding to amino acids 165-180 of human PGRP-S. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG3 κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 188C424 | |
| Unconjugated | |
| PGLYRP1 | |
| Cytokine tag7; MGC126894; MGC126896; peptidoglycan recognition protein 1; Peptidoglycan recognition protein short; Pglyrp; Pglyrp1; PGRP; PGRPS; PGRP-S; SBBI68; Tag7; Tasg7; TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein); TNFSF3L; UNQ639/PRO1269 | |
| Mouse | |
| Protein G | |
| RUO | |
| 21946, 8993 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction