missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGRP-L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561211
This item is not returnable.
View return policy
Description
Immunogen sequence: LMSAPNSGPH NRLYHFLLGA WSLNATELDP CPLSPELLGL TKEVARHDVR EGKEYGVVLA PDGSTVAVEP LLAGLEAGLQ GRRVINLPLD SMAAPWETGD TFPDVVA Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 58%.
Peptidoglycan recognition proteins, such as PGRPL, are part of the innate immune system and recognize peptidoglycan, a ubiquitous component of bacterial cell walls.
Specifications
| PGRP-L | |
| Polyclonal | |
| Unconjugated | |
| PGLYRP2 | |
| C730002N09Rik; HMFT0141; N-acetylmuramoyl-L-alanine amidase; Peptidoglycan recognition protein 2; peptidoglycan recognition protein L; peptidoglycan recognition protein long; PGLYRP2; Pglyrpl; PGRPL; PGRP-L; TagL; tagL-alpha; tagl-beta; TAGL-like; UNQ3103/PRO10102 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114770 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96PD5 | |
| PGLYRP2 | |
| Recombinant protein corresponding to Human PGRP-L. Recombinant protein control fragment (Product #RP-99134). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction